Assay Details
| Gene Symbol: LOC4344497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794776.2 | XP_015650262.1 | 1 - 2 | 331 | 54 | |
| GenBank mRNA | AB110164.1 | - | 1 - 2 | 228 | 54 | |
| AK061391.1 | - | 1 - 2 | 36 | 54 | ||
| AK070120.1 | - | 1 - 2 | 305 | 54 | ||
| AK099353.1 | - | 1 - 2 | 305 | 54 | ||
| Gene Symbol: LOC4345149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287723.1 | - | 2221 | 54 | ||
| Gene Symbol: LOC4352868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288365.1 | - | 2384 | 54 | ||
More Information
Set Membership: |
|

LOC4344497
Genomic Map