Assay Details
Gene Symbol: LOC4344497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794776.2 | XP_015650262.1 | 2 - 3 | 991 | 58 | |
GenBank mRNA | AB110164.1 | - | 2 - 3 | 888 | 58 | |
AK061391.1 | - | 2 - 3 | 696 | 58 | ||
AK070120.1 | - | 2 - 3 | 965 | 58 | ||
AK099353.1 | - | 2 - 3 | 965 | 58 |
Gene Symbol: LOC4345149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK287723.1 | - | 1561 | 58 |
Gene Symbol: LOC4352868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288365.1 | - | 1724 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
