Assay Details
Gene Symbol: LOC4344555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794302.1 | XP_015649788.1 | 2 - 3 | 2234 | 64 | |
GenBank mRNA | AK063293.1 | - | 2 - 3 | 2222 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
