Assay Details
Gene Symbol: LOC4345903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794063.1 | XP_015649549.1 | 1 | 1441 | 74 | |
GenBank mRNA | AK069034.1 | - | 1 | 1448 | 74 | |
AK070422.1 | - | 1 | 1448 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
