Assay Details
| Gene Symbol: LOC4346134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793276.1 | XP_015648762.1 | 7 - 8 | 738 | 74 | |
| GenBank mRNA | AK100263.1 | - | 7 - 8 | 735 | 74 | |
| AY332477.1 | - | 7 - 8 | 717 | 74 | ||
| AY551921.1 | - | 9 - 10 | 1007 | 74 | ||
| U78891.2 | - | 7 - 8 | 767 | 74 | ||
| Y15008.1 | - | 7 - 8 | 671 | 74 | ||
More Information
Set Membership: |
|

LOC4346134
Genomic Map