Assay Details
Gene Symbol: LOC4347886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756282.2 | XP_015611768.1 | 1 - 2 | 302 | 123 | |
GenBank mRNA | AK072521.1 | - | 1 - 2 | 153 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756282.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
