Assay Details
Gene Symbol: LOC4347886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756282.2 | XP_015611768.1 | 2 - 3 | 446 | 75 | |
GenBank mRNA | AK072521.1 | - | 2 - 3 | 297 | 75 | |
AK104879.1 | - | 2 - 3 | 220 | 75 | ||
AY345241.1 | - | 2 - 3 | 200 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
