Assay Details
Gene Symbol: LOC4347886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756282.2 | XP_015611768.1 | 3 - 4 | 504 | 76 | |
GenBank mRNA | AK072521.1 | - | 3 - 4 | 355 | 76 | |
AK104879.1 | - | 3 - 4 | 278 | 76 | ||
AY345241.1 | - | 3 - 4 | 258 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
