Assay Details
Gene Symbol: LOC4347886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756282.2 | XP_015611768.1 | 4 - 5 | 583 | 65 | |
GenBank mRNA | AK072521.1 | - | 4 - 5 | 434 | 65 | |
AK104879.1 | - | 4 - 5 | 357 | 65 | ||
AY345241.1 | - | 4 - 5 | 337 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
