Assay Details
Gene Symbol: LOC4347903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756184.2 | XP_015611670.2 | 5 - 6 | 1004 | 79 | |
GenBank mRNA | AK070300.1 | - | 5 - 6 | 890 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
