Assay Details
Gene Symbol: LOC4347903 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756184.2 | XP_015611670.2 | 6 - 7 | 1264 | 70 | |
GenBank mRNA | AK070300.1 | - | 6 - 7 | 1150 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
