Assay Details
| Gene Symbol: LOC4348176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758770.2 | XP_015614256.1 | 15 - 16 | 1336 | 73 | |
| GenBank mRNA | AK073662.1 | - | 15 - 16 | 1301 | 73 | |
| AK099292.1 | - | 15 - 16 | 1253 | 73 | ||
| AY335488.1 | - | 15 - 16 | 1270 | 73 | ||
| BT129210.1 | - | 4 - 5 | 388 | 73 | ||
| BT129320.1 | - | 4 - 5 | 384 | 73 | ||
| EF122486.1 | - | 15 - 16 | 1287 | 73 | ||
| GQ848037.1 | - | 15 - 16 | 1284 | 73 | ||
| KX302619.1 | - | 15 - 16 | 1275 | 73 | ||
More Information
Set Membership: |
|

LOC4348176
Genomic Map