Assay Details
| Gene Symbol: LOC4348536 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759421.2 | XP_015614907.1 | 6 - 7 | 965 | 113 | |
| XM_015759422.2 | XP_015614908.1 | 6 - 7 | 961 | 113 | ||
| GenBank mRNA | AK068830.1 | - | 7 - 8 | 858 | 113 | |
More Information
Set Membership: |
|

LOC4348536
Genomic Map