Assay Details
| Gene Symbol: LOC4348868 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757761.2 | XP_015613247.1 | 3 - 4 | 2308 | 67 | |
| GenBank mRNA | AK070210.1 | - | 1 - 2 | 41 | 67 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC4348868
Genomic Map