Assay Details
| Gene Symbol: LOC4349071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759559.2 | XP_015615045.1 | 4 - 5 | 874 | 82 | |
| GenBank mRNA | AK061575.1 | - | 4 - 5 | 861 | 82 | |
| AK066499.1 | - | 4 - 5 | 845 | 82 | ||
More Information
Set Membership: |
|

LOC4349071
Genomic Map