Assay Details
| Gene Symbol: LOC4351232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760014.2 | XP_015615500.1 | 1 - 2 | 469 | 151 | |
| GenBank mRNA | AB110185.1 | - | 1 - 2 | 125 | 151 | |
| AK062013.1 | - | 1 - 2 | 143 | 151 | ||
| AK067429.1 | - | 1 - 2 | 265 | 151 | ||
| AK109413.2 | - | 1 - 2 | 261 | 151 | ||
More Information
Set Membership: |
|

LOC4351232
Genomic Map