Assay Details
Gene Symbol: LOC4330142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771558.2 | XP_015627044.1 | 4 - 5 | 776 | 118 | |
GenBank mRNA | AK100979.1 | - | 4 - 5 | 707 | 118 | |
AK242767.1 | - | 4 - 5 | 712 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
