Assay Details
Gene Symbol: LOC4330142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771558.2 | XP_015627044.1 | 7 - 8 | 1029 | 131 | |
GenBank mRNA | AK100979.1 | - | 7 - 8 | 960 | 131 | |
AK242767.1 | - | 7 - 8 | 965 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
