Assay Details
Gene Symbol: LOC4330371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770118.2 | XP_015625604.1 | 17 - 18 | 4864 | 80 | |
XM_015770121.2 | XP_015625607.1 | 4 - 5 | 3403 | 80 | ||
GenBank mRNA | AK120785.1 | - | 2 - 3 | 1517 | 80 | |
GQ906768.1 | - | 16 - 17 | 4631 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
