Assay Details
Gene Symbol: LOC4330371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770118.2 | XP_015625604.1 | 20 - 21 | 6134 | 83 | |
XM_015770121.2 | XP_015625607.1 | 7 - 8 | 4673 | 83 | ||
GenBank mRNA | AK120785.1 | - | 5 - 6 | 2787 | 83 | |
GQ906768.1 | - | 19 - 20 | 5901 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
