Assay Details
| Gene Symbol: LOC4330966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767072.2 | XP_015622558.1 | 11 - 12 | 1728 | 93 | |
| GenBank mRNA | AK060825.1 | - | 7 - 8 | 748 | 93 | |
| AK066747.1 | - | 10 - 11 | 1817 | 93 | ||
More Information
Set Membership: |
|

LOC4330966
Genomic Map