Assay Details
Gene Symbol: LOC4330966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767072.2 | XP_015622558.1 | 3 - 4 | 888 | 145 | |
GenBank mRNA | AK066747.1 | - | 3 - 4 | 877 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
