Assay Details
Gene Symbol: LOC4330966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767072.2 | XP_015622558.1 | 9 - 10 | 1624 | 80 | |
GenBank mRNA | AK060825.1 | - | 5 - 6 | 644 | 80 | |
AK066747.1 | - | 8 - 9 | 1713 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
