Assay Details
Gene Symbol: LOC4330992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767749.2 | XP_015623235.1 | 3 - 4 | 1513 | 55 | |
GenBank mRNA | AK073661.1 | - | 3 - 4 | 1519 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
