Assay Details
| Gene Symbol: LOC4330995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767978.2 | XP_015623464.1 | 9 - 10 | 1059 | 109 | |
| GenBank mRNA | AK065845.1 | - | 10 - 11 | 769 | 109 | |
More Information
Set Membership: |
|

LOC4330995
Genomic Map