Assay Details
| Gene Symbol: LOC4330995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767978.2 | XP_015623464.1 | 2 - 3 | 419 | 71 | |
| XM_015767979.2 | XP_015623465.1 | 2 - 3 | 421 | 71 | ||
| GenBank mRNA | AK065845.1 | - | 3 - 4 | 129 | 71 | |
More Information
Set Membership: |
|

LOC4330995
Genomic Map