Assay Details
| Gene Symbol: LOC4330995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767978.2 | XP_015623464.1 | 4 - 5 | 535 | 99 | |
| XM_015767979.2 | XP_015623465.1 | 4 - 5 | 537 | 99 | ||
| GenBank mRNA | AK065845.1 | - | 5 - 6 | 245 | 99 | |
More Information
Set Membership: |
|

LOC4330995
Genomic Map