Assay Details
Gene Symbol: LOC4331214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771005.2 | XP_015626491.1 | 7 - 8 | 1880 | 101 | |
GenBank mRNA | AK101090.1 | - | 7 - 8 | 1867 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
