Assay Details
| Gene Symbol: LOC4331972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777213.2 | XP_015632699.1 | 2 - 3 | 377 | 65 | |
| GenBank mRNA | AK102337.1 | - | 2 - 3 | 304 | 65 | |
More Information
Set Membership: |
|

LOC4331972
Genomic Map