Assay Details
| Gene Symbol: LOC4331979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773305.2 | XP_015628791.1 | 3 - 4 | 478 | 64 | |
| GenBank mRNA | AK071317.1 | - | 3 - 4 | 485 | 64 | |
More Information
Set Membership: |
|

LOC4331979
Genomic Map