Assay Details
| Gene Symbol: LOC4331979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773305.2 | XP_015628791.1 | 7 - 8 | 992 | 105 | |
| GenBank mRNA | AK071317.1 | - | 7 - 8 | 105 | ||
More Information
Set Membership: |
|

LOC4331979
Genomic Map