Assay Details
| Gene Symbol: LOC4332249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772352.2 | XP_015627838.1 | 3 - 4 | 1612 | 95 | |
| GenBank mRNA | AB117936.1 | - | 3 - 4 | 1012 | 95 | |
| AK243425.1 | - | 3 - 4 | 1119 | 95 | ||
More Information
Set Membership: |
|

LOC4332249
Genomic Map