Assay Details
Gene Symbol: LOC4332285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775632.2 | XP_015631118.1 | 12 - 13 | 1759 | 92 | |
XM_015775634.2 | XP_015631120.1 | 11 - 12 | 1846 | 92 | ||
XM_015775635.2 | XP_015631121.1 | 12 - 13 | 1741 | 92 | ||
GenBank mRNA | AB638270.1 | - | 11 - 12 | 1351 | 92 | |
AK111518.1 | - | 12 - 13 | 1754 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
