Assay Details
Gene Symbol: LOC4332285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775632.2 | XP_015631118.1 | 14 - 15 | 1954 | 96 | |
XM_015775634.2 | XP_015631120.1 | 13 - 14 | 2041 | 96 | ||
XM_015775635.2 | XP_015631121.1 | 14 - 15 | 1936 | 96 | ||
GenBank mRNA | AB638270.1 | - | 13 - 14 | 1546 | 96 | |
AK111518.1 | - | 14 - 15 | 1949 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
