Assay Details
| Gene Symbol: LOC4332285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775632.2 | XP_015631118.1 | 15 - 16 | 2086 | 67 | |
| XM_015775634.2 | XP_015631120.1 | 14 - 15 | 2173 | 67 | ||
| XM_015775635.2 | XP_015631121.1 | 15 - 16 | 2068 | 67 | ||
| GenBank mRNA | AB638270.1 | - | 14 - 15 | 1678 | 67 | |
| AK111518.1 | - | 15 - 16 | 2081 | 67 | ||
More Information
Set Membership: |
|

LOC4332285
Genomic Map