Assay Details
| Gene Symbol: LOC4332285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775632.2 | XP_015631118.1 | 16 - 17 | 2316 | 94 | |
| XM_015775634.2 | XP_015631120.1 | 15 - 16 | 2403 | 94 | ||
| XM_015775635.2 | XP_015631121.1 | 16 - 17 | 2298 | 94 | ||
| GenBank mRNA | AB638270.1 | - | 15 - 16 | 1908 | 94 | |
| AK111518.1 | - | 16 - 17 | 2311 | 94 | ||
More Information
Set Membership: |
|

LOC4332285
Genomic Map