Assay Details
Gene Symbol: LOC4332285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775632.2 | XP_015631118.1 | 20 - 21 | 2844 | 65 | |
XM_015775634.2 | XP_015631120.1 | 19 - 20 | 2931 | 65 | ||
XM_015775635.2 | XP_015631121.1 | 20 - 21 | 2826 | 65 | ||
GenBank mRNA | AB638270.1 | - | 19 - 20 | 2436 | 65 | |
AK111518.1 | - | 20 - 21 | 2839 | 65 | ||
AK112052.1 | - | 1 - 2 | 47 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
