Assay Details
| Gene Symbol: LOC4332285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775632.2 | XP_015631118.1 | 21 - 22 | 3019 | 73 | |
| XM_015775634.2 | XP_015631120.1 | 20 - 21 | 3106 | 73 | ||
| XM_015775635.2 | XP_015631121.1 | 21 - 22 | 3001 | 73 | ||
| GenBank mRNA | AB638270.1 | - | 20 - 21 | 2611 | 73 | |
| AK111518.1 | - | 21 - 22 | 3014 | 73 | ||
| AK112052.1 | - | 2 - 3 | 222 | 73 | ||
More Information
Set Membership: |
|

LOC4332285
Genomic Map