Assay Details
| Gene Symbol: LOC4332285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775632.2 | XP_015631118.1 | 23 - 24 | 3248 | 104 | |
| XM_015775634.2 | XP_015631120.1 | 22 - 23 | 3335 | 104 | ||
| XM_015775635.2 | XP_015631121.1 | 23 - 24 | 3230 | 104 | ||
| GenBank mRNA | AB638270.1 | - | 22 - 23 | 2840 | 104 | |
| AK111518.1 | - | 23 - 24 | 3243 | 104 | ||
| AK112052.1 | - | 4 - 5 | 451 | 104 | ||
More Information
Set Membership: |
|

LOC4332285
Genomic Map