Assay Details
Gene Symbol: LOC4332285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775632.2 | XP_015631118.1 | 24 - 25 | 3434 | 93 | |
XM_015775634.2 | XP_015631120.1 | 23 - 24 | 3521 | 93 | ||
XM_015775635.2 | XP_015631121.1 | 24 - 25 | 3416 | 93 | ||
GenBank mRNA | AB638270.1 | - | 23 - 24 | 3026 | 93 | |
AK111518.1 | - | 24 - 25 | 3429 | 93 | ||
AK112052.1 | - | 5 - 6 | 637 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
