Assay Details
Gene Symbol: LOC4332285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775632.2 | XP_015631118.1 | 25 - 26 | 3604 | 75 | |
XM_015775634.2 | XP_015631120.1 | 24 - 25 | 3691 | 75 | ||
XM_015775635.2 | XP_015631121.1 | 25 - 26 | 3586 | 75 | ||
GenBank mRNA | AB638270.1 | - | 24 - 25 | 3196 | 75 | |
AK111518.1 | - | 25 - 26 | 3599 | 75 | ||
AK112052.1 | - | 6 - 7 | 807 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
