Assay Details
| Gene Symbol: LOC4332285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775632.2 | XP_015631118.1 | 6 - 7 | 1121 | 98 | |
| XM_015775634.2 | XP_015631120.1 | 5 - 6 | 1208 | 98 | ||
| XM_015775635.2 | XP_015631121.1 | 6 - 7 | 1121 | 98 | ||
| GenBank mRNA | AB638270.1 | - | 5 - 6 | 713 | 98 | |
| AK111518.1 | - | 6 - 7 | 1116 | 98 | ||
More Information
Set Membership: |
|

LOC4332285
Genomic Map