Assay Details
| Gene Symbol: LOC4333310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772387.2 | XP_015627873.1 | 1 - 2 | 169 | 118 | |
| GenBank mRNA | AK061735.1 | - | 1 - 2 | 140 | 118 | |
| AK098953.1 | - | 1 - 2 | 146 | 118 | ||
| AK099558.1 | - | 1 - 2 | 153 | 118 | ||
| AK104547.1 | - | 1 - 2 | 136 | 118 | ||
| AK242951.1 | - | 2591 | 118 | |||
| AK289006.1 | - | 2591 | 118 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772387.2 transcripts.
Set Membership: |
|

LOC4333310
Genomic Map