Assay Details
| Gene Symbol: LOC4333310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772387.2 | XP_015627873.1 | 2 - 3 | 221 | 70 | |
| GenBank mRNA | AK061735.1 | - | 2 - 3 | 192 | 70 | |
| AK098953.1 | - | 2 - 3 | 198 | 70 | ||
| AK099558.1 | - | 2 - 3 | 205 | 70 | ||
| AK104547.1 | - | 2 - 3 | 188 | 70 | ||
| AK242951.1 | - | 2643 | 70 | |||
| AK289006.1 | - | 2643 | 70 | |||
| BT132418.1 | - | 2 - 3 | 82 | 70 | ||
More Information
Set Membership: |
|

LOC4333310
Genomic Map