Assay Details
| Gene Symbol: LOC4333310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772387.2 | XP_015627873.1 | 3 - 4 | 323 | 77 | |
| GenBank mRNA | AK061735.1 | - | 3 - 4 | 294 | 77 | |
| AK098953.1 | - | 3 - 4 | 300 | 77 | ||
| AK099558.1 | - | 3 - 4 | 307 | 77 | ||
| AK104547.1 | - | 3 - 4 | 290 | 77 | ||
| AK242951.1 | - | 2745 | 77 | |||
| AK289006.1 | - | 2745 | 77 | |||
| BT132418.1 | - | 3 - 4 | 184 | 77 | ||
More Information
Set Membership: |
|

LOC4333310
Genomic Map