Assay Details
| Gene Symbol: LOC4333310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772387.2 | XP_015627873.1 | 6 - 7 | 714 | 76 | |
| GenBank mRNA | AK061735.1 | - | 6 - 7 | 685 | 76 | |
| AK098953.1 | - | 6 - 7 | 691 | 76 | ||
| AK099558.1 | - | 6 - 7 | 698 | 76 | ||
| AK104547.1 | - | 6 - 7 | 681 | 76 | ||
| AK242951.1 | - | 3136 | 76 | |||
| AK289006.1 | - | 3136 | 76 | |||
| BT132418.1 | - | 6 - 7 | 575 | 76 | ||
More Information
Set Membership: |
|

LOC4333310
Genomic Map