Assay Details
Gene Symbol: LOC4333349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776295.2 | XP_015631781.1 | 12 - 13 | 2389 | 111 | |
GenBank mRNA | AK121995.1 | - | 12 - 13 | 2392 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
