Assay Details
Gene Symbol: LOC4333349 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776295.2 | XP_015631781.1 | 2 - 3 | 549 | 58 | |
GenBank mRNA | AK121995.1 | - | 2 - 3 | 548 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
