Assay Details
Gene Symbol: LOC9271868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772830.2 | XP_015628316.1 | 1 - 2 | 279 | 133 | |
GenBank mRNA | AK106686.1 | - | 1 - 2 | 254 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772830.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
