Assay Details
Gene Symbol: LOC9271868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772830.2 | XP_015628316.1 | 3 - 4 | 513 | 102 | |
GenBank mRNA | AK106686.1 | - | 3 - 4 | 488 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
